Assay Details
Gene Symbol: PAPSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015880.1 | NP_001015880.1 | 9 - 10 | 1353 | 96 | |
NM_004670.3 | NP_004661.2 | 8 - 9 | 1338 | 96 | ||
GenBank mRNA | AF074331.1 | - | 8 - 9 | 1138 | 96 | |
AF091242.1 | - | 8 - 9 | 1155 | 96 | ||
AF150754.2 | - | 9 - 10 | 1170 | 96 | ||
AF173365.1 | - | 9 - 10 | 1138 | 96 | ||
AK301453.1 | - | 8 - 9 | 1125 | 96 | ||
AK312556.1 | - | 8 - 9 | 1179 | 96 | ||
BC009894.2 | - | 8 - 9 | 1186 | 2988345 | 96 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000621.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
sulfate assimilation
skeletal system development
blood coagulation
phosphorylation
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
bone development
adenylylsulfate kinase activity
sulfate adenylyltransferase (ATP) activity
ATP binding
nucleotidyltransferase activity
sulfate assimilation
skeletal system development
blood coagulation
phosphorylation
3'-phosphoadenosine 5'-phosphosulfate biosynthetic process
bone development
adenylylsulfate kinase activity
sulfate adenylyltransferase (ATP) activity
ATP binding
nucleotidyltransferase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | ENST00000361175.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | ENST00000371981.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | ENST00000427144.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10006821.hg.1 | TC10000621.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000156598.hg.1 | TC1000008354.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000361175.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000456849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000465996.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | ENST00000482258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_001015880.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000008354.hg.1 | NM_004670.3 | Refseq |
