Assay Details
| Gene Symbol: GUCY2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004963.3 | NP_004954.2 | 5 - 6 | 868 | 108 | |
| XM_011520631.2 | XP_011518933.1 | 5 - 6 | 766 | 108 | ||
| GenBank mRNA | AK310034.1 | - | 5 - 6 | 858 | 108 | |
| BC136544.1 | - | 5 - 6 | 777 | 9020534 | 108 | |
| BC136545.1 | - | 5 - 6 | 777 | 9020535 | 108 | |
| M73489.1 | - | 5 - 6 | 779 | 108 | ||
| S57551.1 | - | 5 - 6 | 848 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001271.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cGMP biosynthetic process
protein phosphorylation
receptor guanylyl cyclase signaling pathway
response to toxic substance
intracellular signal transduction
regulation of cell proliferation
adenylate cyclase activity
guanylate cyclase activity
protein kinase activity
protein binding
ATP binding
GTP binding
toxic substance binding
cGMP biosynthetic process
protein phosphorylation
receptor guanylyl cyclase signaling pathway
response to toxic substance
intracellular signal transduction
regulation of cell proliferation
adenylate cyclase activity
guanylate cyclase activity
protein kinase activity
protein binding
ATP binding
GTP binding
toxic substance binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12016556.hg.1 | ENST00000261170.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016556.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12016556.hg.1 | TC12001271.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200175578.hg.1 | TC1200010009.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000261170.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | ENST00000535803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010009.hg.1 | NM_004963.3 | Refseq |

GUCY2C
Genomic Map