Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002755.hg.1 |
ENST00000268711.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002755.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002755.hg.1 |
TC17000199.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000580462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000580462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000580462.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000581315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000581315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000581315.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000585041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000585041.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149854.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149854.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149854.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149854.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149854.hg.1 |
TC1700007068.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000580462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000580462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000580462.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000581315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000581315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000581315.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000585041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000585041.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
ENST00000624097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007068.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149855.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149855.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149855.hg.1 |
ENST00000268711.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149855.hg.1 |
NM_018019.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149855.hg.1 |
TC1700007068.hg.1 |
Clariom D TC |