Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198546.1 | NP_940948.1 | 12 - 13 | 1838 | 62 | |
| XM_011541407.2 | XP_011539709.1 | 13 - 14 | 2350 | 62 | ||
| XM_017001224.1 | XP_016856713.1 | 9 - 10 | 1398 | 62 | ||
| GenBank mRNA | AY327405.1 | - | 12 - 13 | 1838 | 62 | |
| BC150598.1 | - | 10 - 11 | 1620 | 9056968 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002263.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01034538.hg.1 | TC01002263.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034538.hg.1 | ENST00000335496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034538.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01034538.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196141.hg.1 | TC0100013042.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100196140.hg.1 | TC0100013042.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 208776 | - | - |

SPATA21
Genomic Map