Assay Details
Gene Symbol: SPG20-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045180.1 | 4 - 5 | 385 | 85 | ||
NR_045181.1 | 3 - 4 | 298 | 85 | |||
GenBank mRNA | AW292824.1 | - | 4 - 5 | 384 | 85 | |
BF732820.1 | - | 3 - 4 | 298 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC13000123.hg.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR13001390.hg.1 | ENST00000379848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001390.hg.1 | ENST00000493739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001390.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001390.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001390.hg.1 | TC13000123.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR13001389.hg.1 | ENST00000379848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001389.hg.1 | ENST00000493739.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR13001389.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001389.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR13001389.hg.1 | TC13000123.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146759.hg.1 | TC1300006877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1300146757.hg.1 | TC1300006877.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000379848.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000488319.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | ENST00000493739.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045180.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1300006877.hg.1 | NR_045181.1 | Refseq |
