Assay Details
| Gene Symbol: CD34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025109.1 | NP_001020280.1 | 2 - 3 | 520 | 151 | |
| NM_001773.2 | NP_001764.1 | 2 - 3 | 520 | 151 | ||
| GenBank mRNA | AF523361.1 | - | 2 - 3 | 333 | 151 | |
| AF538601.1 | - | 1 - 2 | 171 | 151 | ||
| AK291529.1 | - | 2 - 3 | 273 | 151 | ||
| AK293030.1 | - | 2 - 3 | 297 | 151 | ||
| AK300835.1 | - | 3 - 4 | 337 | 151 | ||
| BC039146.1 | - | 2 - 3 | 270 | 4746591 | 151 | |
| M81104.1 | - | 2 - 3 | 519 | 151 | ||
| S53910.1 | - | 2 - 3 | 352 | 151 | ||
| S53911.1 | - | 2 - 3 | 352 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003777.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
tissue homeostasis
endothelial cell proliferation
glomerular filtration
endothelium development
cell-matrix adhesion
signal transduction
regulation of blood pressure
cell proliferation
positive regulation of gene expression
negative regulation of gene expression
single organismal cell-cell adhesion
hemopoiesis
negative regulation of blood coagulation
negative regulation of tumor necrosis factor production
positive regulation of interleukin-10 production
mesangial cell-matrix adhesion
paracrine signaling
positive regulation of odontogenesis
negative regulation of nitric oxide biosynthetic process
positive regulation of angiogenesis
cell motility
regulation of immune response
leukocyte migration
transdifferentiation
vascular wound healing
hematopoietic stem cell proliferation
positive regulation of transforming growth factor beta production
positive regulation of granulocyte colony-stimulating factor production
extracellular exosome assembly
glomerular endothelium development
stem cell proliferation
metanephric glomerular mesangial cell differentiation
negative regulation of cellular response to heat
negative regulation of cellular response to hypoxia
negative regulation of interleukin-2 secretion
positive regulation of glial cell-derived neurotrophic factor secretion
negative regulation of neuron death
positive regulation of vasculogenesis
transcription factor binding
carbohydrate binding
sulfate binding
tissue homeostasis
endothelial cell proliferation
glomerular filtration
endothelium development
cell-matrix adhesion
signal transduction
regulation of blood pressure
cell proliferation
positive regulation of gene expression
negative regulation of gene expression
single organismal cell-cell adhesion
hemopoiesis
negative regulation of blood coagulation
negative regulation of tumor necrosis factor production
positive regulation of interleukin-10 production
mesangial cell-matrix adhesion
paracrine signaling
positive regulation of odontogenesis
negative regulation of nitric oxide biosynthetic process
positive regulation of angiogenesis
cell motility
regulation of immune response
leukocyte migration
transdifferentiation
vascular wound healing
hematopoietic stem cell proliferation
positive regulation of transforming growth factor beta production
positive regulation of granulocyte colony-stimulating factor production
extracellular exosome assembly
glomerular endothelium development
stem cell proliferation
metanephric glomerular mesangial cell differentiation
negative regulation of cellular response to heat
negative regulation of cellular response to hypoxia
negative regulation of interleukin-2 secretion
positive regulation of glial cell-derived neurotrophic factor secretion
negative regulation of neuron death
positive regulation of vasculogenesis
transcription factor binding
carbohydrate binding
sulfate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01057879.hg.1 | TC01003777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057879.hg.1 | ENST00000310833.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057879.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057879.hg.1 | ENST00000367037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057879.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057879.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | TC01003777.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | ENST00000310833.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | ENST00000367037.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057878.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229966.hg.1 | TC0100017142.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000367036.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017142.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | ENST00000310833.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | ENST00000356522.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | ENST00000485761.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | NM_001025109.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | NM_001773.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229965.hg.1 | TC0100017142.hg.1 | Clariom D TC |

CD34
Genomic Map