Assay Details
Gene Symbol: NOTUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178493.5 | NP_848588.3 | 1 - 2 | 595 | 59 | |
GenBank mRNA | BC036872.1 | - | 1 - 2 | 144 | 5225889 | 59 |
BC060882.1 | - | 1 - 2 | 603 | 30336041 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001968.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17026839.hg.1 | ENST00000409678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026839.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026839.hg.1 | TC17001968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026842.hg.1 | ENST00000409678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026842.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026842.hg.1 | TC17001968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026840.hg.1 | ENST00000409678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026840.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026840.hg.1 | TC17001968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026841.hg.1 | ENST00000409678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026841.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026841.hg.1 | TC17001968.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026837.hg.1 | ENST00000409678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026837.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026837.hg.1 | TC17001968.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199078.hg.1 | TC1700012084.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199079.hg.1 | TC1700012084.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199076.hg.1 | TC1700012084.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700199082.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199082.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199082.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199082.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199082.hg.1 | TC1700012084.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199080.hg.1 | TC1700012084.hg.1 | Clariom D TC |
