Assay Details
Gene Symbol: NOTUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178493.5 | NP_848588.3 | 6 - 7 | 972 | 106 | |
GenBank mRNA | AB062438.1 | - | 6 - 7 | 398 | 106 | |
BC060882.1 | - | 6 - 7 | 980 | 30336041 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001968.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026831.hg.1 | ENST00000409678.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026831.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026831.hg.1 | TC17001968.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | ENST00000489218.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012084.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | ENST00000409678.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | ENST00000425009.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | ENST00000477214.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | NM_178493.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700199070.hg.1 | TC1700012084.hg.1 | Clariom D TC |
