Assay Details
Gene Symbol: TBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004608.3 | NP_004599.2 | 4 - 5 | 827 | 75 | |
XM_005255523.2 | XP_005255580.1 | 5 - 6 | 1112 | 75 | ||
XM_011545926.2 | XP_011544228.1 | 5 - 6 | 1275 | 75 | ||
XM_017023614.1 | XP_016879103.1 | 5 - 6 | 852 | 75 | ||
GenBank mRNA | AJ007989.1 | - | 4 - 5 | 791 | 75 | |
AK022330.1 | - | 4 - 5 | 1461 | 75 | ||
BC026031.1 | - | 4 - 5 | 827 | 5460190 | 75 | |
KJ534981.1 | - | 4 - 5 | 863 | 75 | ||
KJ535035.1 | - | 3 - 4 | 870 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001018.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
mesoderm formation
cell fate specification
transcription, DNA-templated
mesoderm development
anatomical structure morphogenesis
negative regulation of neuron projection development
negative regulation of neuron maturation
signal transduction involved in regulation of gene expression
somite rostral/caudal axis specification
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
mesoderm formation
cell fate specification
transcription, DNA-templated
mesoderm development
anatomical structure morphogenesis
negative regulation of neuron projection development
negative regulation of neuron maturation
signal transduction involved in regulation of gene expression
somite rostral/caudal axis specification
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II transcription factor binding
RNA polymerase II activating transcription factor binding
transcriptional repressor activity, RNA polymerase II transcription factor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16014561.hg.1 | ENST00000279386.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16014561.hg.1 | ENST00000395224.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16014561.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16014561.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16014561.hg.1 | TC16001018.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | ENST00000627355.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009951.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000279386.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000395224.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000553607.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | ENST00000567664.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | NM_004608.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600176697.hg.1 | TC1600009951.hg.1 | Clariom D TC |
