Assay Details
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014681.5 | NP_055496.2 | 15 - 16 | 3509 | 69 | |
XM_005259443.3 | XP_005259500.1 | 15 - 16 | 4020 | 69 | ||
XM_011527550.2 | XP_011525852.1 | 15 - 16 | 4144 | 69 | ||
XM_011527551.2 | XP_011525853.1 | 15 - 16 | 3458 | 69 | ||
XM_017027524.1 | XP_016883013.1 | 14 - 15 | 3423 | 69 | ||
GenBank mRNA | AK297692.1 | - | 10 - 11 | 1668 | 69 | |
BQ643547.1 | - | 2 - 3 | 227 | 69 | ||
D50924.1 | - | 10 - 11 | 2560 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000678.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009495.hg.1 | ENST00000257252.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009495.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009495.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009495.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164735.hg.1 | NM_014681.5 | Refseq |
