Assay Details
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D50924.1 | - | 2 - 3 | 1305 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
