Assay Details
| Gene Symbol: MUC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173600.2 | NP_775871.2 | 24499 | 68 | ||
| GenBank mRNA | AK093065.1 | - | 28 - 29 | 1491 | 68 | |
| AY236870.1 | - | 27 - 28 | 1397 | 68 | ||
| HM801842.1 | - | 24499 | 68 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat kinase 2 | LRRK2 |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
-192.3kb | Sense | Chr. 12 - 40224895 - 40369285 on NCBI Build GRCh38 |
| Downstream | contactin 1 | CNTN1 |
F3, GP135, MYPCN |
128.9kb | Sense | Chr. 12 - 40692439 - 41072412 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

MUC19
Genomic Map