Assay Details
| Gene Symbol: MUC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173600.2 | NP_775871.2 | 24565 | 110 | ||
| GenBank mRNA | AK093065.1 | - | 30 - 31 | 1558 | 110 | |
| AY236870.1 | - | 29 - 30 | 1463 | 110 | ||
| HM801842.1 | - | 24565 | 110 | |||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DN831808.1 | - | Chr. - - [-] on Build | 110 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat kinase 2 | LRRK2 |
AURA17, DARDARIN, PARK8, RIPK7, ROCO2 |
-196.2kb | Sense | Chr. 12 - 40224895 - 40369285 on NCBI Build GRCh38 |
| Downstream | contactin 1 | CNTN1 |
F3, GP135, MYPCN |
126.6kb | Sense | Chr. 12 - 40692439 - 41072412 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

MUC19
Genomic Map