Assay Details
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014681.5 | NP_055496.2 | 1 - 2 | 78 | 75 | |
GenBank mRNA | BM543601.1 | - | 1 - 2 | 70 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000678.hg
Additional Information:
This assay is in the 5'UTR region of the NM_014681.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19009465.hg.1 | ENST00000257252.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009465.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009465.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009465.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009469.hg.1 | ENST00000257252.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009469.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009469.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009469.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19009468.hg.1 | ENST00000257252.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009468.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009468.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009468.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1900164707.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164707.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164707.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164707.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164707.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164705.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164705.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164705.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164705.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164705.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164704.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164704.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164704.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164704.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164704.hg.1 | NM_014681.5 | Refseq |
