Assay Details
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014681.5 | NP_055496.2 | 3 - 4 | 1365 | 106 | |
XM_005259443.3 | XP_005259500.1 | 3 - 4 | 1876 | 106 | ||
XM_011527550.2 | XP_011525852.1 | 3 - 4 | 2000 | 106 | ||
XM_011527551.2 | XP_011525853.1 | 3 - 4 | 1314 | 106 | ||
XM_017027524.1 | XP_016883013.1 | 3 - 4 | 1497 | 106 | ||
GenBank mRNA | AK304710.1 | - | 3 - 4 | 1328 | 106 | |
BQ708034.1 | - | 1 - 2 | 180 | 106 | ||
DR762320.1 | - | 2 - 3 | 733 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000678.hg
Additional Information:
This assay is in the 5'UTR region of the XM_017027524.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19009471.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009471.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009471.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164710.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164710.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164710.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164710.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164710.hg.1 | NM_014681.5 | Refseq |
