Assay Details
| Gene Symbol: RGS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286673.1 | NP_001273602.1 | 1 - 2 | 255 | 58 | |
| GenBank mRNA | AF366054.1 | - | 1 - 2 | 163 | 58 | |
More Information
Set Membership: |
|

RGS20
Genomic Map