Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015399.3 | NP_056214.1 | 8 - 9 | 843 | 93 | |
| GenBank mRNA | AF147350.1 | - | 2 - 3 | 157 | 93 | |
| AF159141.1 | - | 8 - 9 | 817 | 93 | ||
| AK313773.1 | - | 8 - 9 | 926 | 93 | ||
| BC009834.2 | - | 8 - 9 | 799 | 3959486 | 93 | |
| CR457173.1 | - | 7 - 8 | 696 | 93 | ||
More Information
Set Membership: |
|

BRMS1
Genomic Map