Assay Details
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161580.1 | NP_001155052.1 | 9 - 10 | 1303 | 75 | |
XM_017006104.1 | XP_016861593.1 | 9 - 10 | 1096 | 75 | ||
GenBank mRNA | BC110877.1 | - | 9 - 10 | 1303 | 5588109 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001447.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03026158.hg.1 | TC03001447.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026158.hg.1 | ENST00000296484.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026158.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026158.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026158.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026159.hg.1 | TC03001447.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026159.hg.1 | ENST00000296484.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026159.hg.1 | ENST00000394970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026159.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026159.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026159.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183215.hg.1 | TC0300011154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183213.hg.1 | TC0300011154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183214.hg.1 | TC0300011154.hg.1 | Clariom D TC |
