Assay Details
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161580.1 | NP_001155052.1 | 3 - 4 | 591 | 76 | |
NM_001161581.1 | NP_001155053.1 | 3 - 4 | 662 | 76 | ||
NM_015426.4 | NP_056241.3 | 3 - 4 | 591 | 76 | ||
XM_011533560.1 | XP_011531862.1 | 3 - 4 | 313 | 76 | ||
XM_011533561.1 | XP_011531863.1 | 3 - 4 | 313 | 76 | ||
XM_011533562.2 | XP_011531864.1 | 3 - 4 | 313 | 76 | ||
XM_011533563.1 | XP_011531865.1 | 3 - 4 | 313 | 76 | ||
XM_011533564.2 | XP_011531866.1 | 3 - 4 | 313 | 76 | ||
XM_011533565.2 | XP_011531867.1 | 3 - 4 | 313 | 76 | ||
XM_017006104.1 | XP_016861593.1 | 3 - 4 | 384 | 76 | ||
GenBank mRNA | AK075289.1 | - | 3 - 4 | 306 | 76 | |
AK315688.1 | - | 3 - 4 | 313 | 76 | ||
BC007417.2 | - | 2 - 3 | 241 | 76 | ||
BC110877.1 | - | 3 - 4 | 591 | 5588109 | 76 | |
BC119692.2 | - | 3 - 4 | 301 | 40116721 | 76 | |
BC119693.2 | - | 3 - 4 | 377 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001447.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | TC03001447.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | ENST00000296484.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | ENST00000394970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026167.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026170.hg.1 | TC03001447.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026170.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026170.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183223.hg.1 | TC0300011154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183227.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183227.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183227.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183227.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183227.hg.1 | TC0300011154.hg.1 | Clariom D TC |
