Assay Details
Gene Symbol: POC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161580.1 | NP_001155052.1 | 6 - 7 | 997 | 72 | |
NM_001161581.1 | NP_001155053.1 | 6 - 7 | 1068 | 72 | ||
NM_015426.4 | NP_056241.3 | 6 - 7 | 997 | 72 | ||
XM_011533560.1 | XP_011531862.1 | 6 - 7 | 719 | 72 | ||
XM_011533561.1 | XP_011531863.1 | 6 - 7 | 719 | 72 | ||
XM_011533562.2 | XP_011531864.1 | 6 - 7 | 719 | 72 | ||
XM_017006104.1 | XP_016861593.1 | 6 - 7 | 790 | 72 | ||
GenBank mRNA | AK315688.1 | - | 6 - 7 | 719 | 72 | |
BC007417.2 | - | 5 - 6 | 647 | 72 | ||
BC110877.1 | - | 6 - 7 | 997 | 5588109 | 72 | |
BC119692.2 | - | 6 - 7 | 707 | 40116721 | 72 | |
BC119693.2 | - | 6 - 7 | 783 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001447.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | TC03001447.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | ENST00000296484.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | ENST00000394970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026163.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | TC03001447.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | ENST00000296484.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | ENST00000394970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03026164.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183219.hg.1 | TC0300011154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300183220.hg.1 | TC0300011154.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000296484.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000394970.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | ENST00000474012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161580.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_001161581.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011154.hg.1 | NM_015426.4 | Refseq |
