Assay Details
| Gene Symbol: ASCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005261775.2 | XP_005261832.1 | 1 - 2 | 103 | 130 | |
| XM_011530442.2 | XP_011528744.1 | 1 - 2 | 103 | 130 | ||
| XM_011530445.2 | XP_011528747.1 | 1 - 2 | 103 | 130 | ||
| XM_011530454.2 | XP_011528756.1 | 1 - 2 | 103 | 130 | ||
| XM_017029000.1 | XP_016884489.1 | 1 - 2 | 103 | 130 | ||
| XM_017029008.1 | XP_016884497.1 | 1 - 2 | 103 | 130 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005261775.2, XM_011530442.2, XM_011530445.2, XM_011530454.2, XM_017029000.1, XM_017029008.1 transcripts.
Set Membership: |
|

ASCC2
Genomic Map