Assay Details
| Gene Symbol: PRPF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322267.1 | NP_001309196.1 | 1 - 2 | 142 | 65 | |
| NM_004697.4 | NP_004688.2 | 1 - 2 | 142 | 65 | ||
| NR_136265.1 | 1 - 2 | 142 | 65 | |||
| GenBank mRNA | AF016369.1 | - | 1 - 2 | 92 | 65 | |
| AK313131.1 | - | 1 - 2 | 84 | 65 | ||
| BC001588.2 | - | 1 - 2 | 82 | 2988225 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AF016369.1,AK313131.1,BC001588.2,NM_001322267.1,NM_004697.4 transcripts.
This assay is in the 5'UTR region of the AF016369.1,AK313131.1,BC001588.2,NM_001322267.1,NM_004697.4 transcripts.
Set Membership: |
|

PRPF4
Genomic Map