Assay Details
Gene Symbol: NRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013983.2 | NP_053586.1 | 6 - 7 | 1422 | 71 | |
XM_006714810.3 | XP_006714873.1 | 7 - 8 | 1865 | 71 | ||
XM_017010065.1 | XP_016865554.1 | 7 - 8 | 748 | 71 | ||
GenBank mRNA | AK124504.1 | - | 8 - 9 | 1428 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
