Assay Details
| Gene Symbol: NDUFV2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021074.4 | NP_066552.2 | 3 - 4 | 302 | 112 | |
| XM_017025782.1 | XP_016881271.1 | 3 - 4 | 492 | 112 | ||
| GenBank mRNA | AK291865.1 | - | 3 - 4 | 198 | 112 | |
| AK312129.1 | - | 3 - 4 | 220 | 112 | ||
| BC001632.1 | - | 3 - 4 | 212 | 3536345 | 112 | |
| BC017487.1 | - | 3 - 4 | 210 | 3537815 | 112 | |
| BC071689.1 | - | 3 - 4 | 192 | 4372795 | 112 | |
| BG742948.1 | - | 3 - 4 | 302 | 112 | ||
| CB121843.1 | - | 3 - 4 | 255 | 112 | ||
| CR456928.1 | - | 3 - 4 | 188 | 112 | ||
| M22538.1 | - | 3 - 4 | 206 | 112 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000041.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial electron transport, NADH to ubiquinone
nervous system development
mitochondrial respiratory chain complex I assembly
cardiac muscle tissue development
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding
mitochondrial electron transport, NADH to ubiquinone
nervous system development
mitochondrial respiratory chain complex I assembly
cardiac muscle tissue development
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18000429.hg.1 | ENST00000318388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000429.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000429.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000429.hg.1 | TC18000041.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000430.hg.1 | ENST00000318388.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000430.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000430.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18000430.hg.1 | TC18000041.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | TC1800009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163200.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000474740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000497577.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000577703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | ENST00000583375.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800009214.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | NM_021074.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | TC1800009214.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000318388.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000400033.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000465096.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000474350.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | ENST00000483511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800163199.hg.1 | NM_021074.4 | Refseq |

NDUFV2
Genomic Map