Assay Details
Gene Symbol: DHX34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014681.5 | NP_055496.2 | 11 - 12 | 2828 | 61 | |
XM_005259443.3 | XP_005259500.1 | 11 - 12 | 3339 | 61 | ||
XM_011527550.2 | XP_011525852.1 | 11 - 12 | 3463 | 61 | ||
XM_011527551.2 | XP_011525853.1 | 11 - 12 | 2777 | 61 | ||
XM_017027524.1 | XP_016883013.1 | 10 - 11 | 2742 | 61 | ||
GenBank mRNA | AA454494.1 | - | 3 - 4 | 61 | ||
AK297692.1 | - | 6 - 7 | 987 | 61 | ||
BF894697.1 | - | 1 - 2 | 184 | 61 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000678.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19009483.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009483.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009483.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19009485.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009485.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009485.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR19009486.hg.1 | ENST00000257252.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009486.hg.1 | ENST00000328771.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19009486.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19009486.hg.1 | TC19000678.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164727.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000471451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | TC1900008420.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | NM_014681.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | TC1900008420.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | ENST00000328771.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | ENST00000460681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | ENST00000486327.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900164725.hg.1 | NM_014681.5 | Refseq |
