Assay Details
| Gene Symbol: CNOT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256741.1 | NP_001243670.1 | 12 - 13 | 1829 | 89 | |
| NR_046352.1 | 11 - 12 | 1667 | 89 | |||
| XM_011533566.2 | XP_011531868.1 | 12 - 13 | 1785 | 89 | ||
| XM_017006112.1 | XP_016861601.1 | 12 - 13 | 1829 | 89 | ||
| GenBank mRNA | AK021695.1 | - | 10 - 11 | 1263 | 89 | |
| AK309670.1 | - | 11 - 12 | 1592 | 89 | ||
| BC002928.2 | - | 12 - 13 | 1731 | 3954507 | 89 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CNOT10
Genomic Map