Assay Details
Gene Symbol: OXSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005109.2 | NP_005100.1 | 16 - 17 | 1787 | 72 | |
XM_005265638.2 | XP_005265695.1 | 17 - 18 | 1928 | 72 | ||
XM_011534331.2 | XP_011532633.1 | 16 - 17 | 1839 | 72 | ||
XM_017007601.1 | XP_016863090.1 | 15 - 16 | 1594 | 72 | ||
XM_017007602.1 | XP_016863091.1 | 18 - 19 | 2114 | 72 | ||
GenBank mRNA | AB017642.1 | - | 16 - 17 | 1787 | 72 | |
AB029024.1 | - | 16 - 17 | 1540 | 72 | ||
BC008726.2 | - | 16 - 17 | 1770 | 3163379 | 72 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000188.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
response to oxidative stress
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
intracellular signal transduction
cellular hypotonic response
negative regulation of potassium ion transmembrane transporter activity
negative regulation of rubidium ion transmembrane transporter activity
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
protein phosphorylation
response to oxidative stress
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
intracellular signal transduction
cellular hypotonic response
negative regulation of potassium ion transmembrane transporter activity
negative regulation of rubidium ion transmembrane transporter activity
magnesium ion binding
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000492714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | ENST00000492714.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03003656.hg.1 | TC03000188.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03003656.hg.1 | ENST00000311806.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03003656.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000426620.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000446845.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000467900.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000483695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000492714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | ENST00000492714.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007068.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300149460.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300149460.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300149460.hg.1 | ENST00000311806.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300149460.hg.1 | NM_005109.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300149460.hg.1 | TC0300007068.hg.1 | Clariom D TC |
