Assay Details
Gene Symbol: ST20-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028330.1 | 1 | 2029 | 121 | ||
GenBank mRNA | AK091107.1 | - | 1 | 2029 | 121 | |
AK092679.1 | - | 1 | 1635 | 121 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P85330 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1568 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 11671 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 12510 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P85330 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_32_P85330 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279248 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279203 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781496 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16812562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279198 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781457 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781486 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781466 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781469 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279199 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279238 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279225 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781456 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279235 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781494 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781480 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279198 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279190 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781469 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279201 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781503 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16812562 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781457 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279248 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279199 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781466 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781482 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781455 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781491 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279235 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279196 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781504 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781492 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279192 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781495 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279225 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279203 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781456 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30279238 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781496 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09781486 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P85330 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P360387 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 7985254 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7985253 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7990815 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1556588_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11735074_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11735074_at | - | - |
Affymetrix Human Genome U219 Array | 11735075_at | - | - |
Affymetrix Human Genome U219 Array | 11735075_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1556588_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1556588_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1556588_at6 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053143 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053143 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 229882 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1768855 | CR600752.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768855 | CR616297.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768855 | AK092679.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768855 | BC016975.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768855 | NR_028330.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768855 | CR600752.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768855 | CR616297.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768855 | AK092679.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768855 | BC016975.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768855 | NR_028330.1 | human_transcriptome |
