Assay Details
Gene Symbol: ABCB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003742.2 | NP_003733.2 | 3 - 4 | 216 | 93 | |
XM_006712817.3 | XP_006712880.1 | 3 - 4 | 328 | 135 | ||
XM_011512077.2 | XP_011510379.1 | 3 - 4 | 388 | 195 | ||
XM_011512078.2 | XP_011510380.1 | 3 - 4 | 388 | 195 | ||
XM_011512080.2 | XP_011510382.1 | 3 - 4 | 391 | 195 | ||
XM_017005165.1 | XP_016860654.1 | 3 - 4 | 390 | 195 | ||
GenBank mRNA | AF091582.1 | - | 3 - 4 | 216 | 93 | |
AF136523.1 | - | 3 - 4 | 217 | 93 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002508.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
bile acid biosynthetic process
transport
bile acid and bile salt transport
canalicular bile acid transport
sodium ion transmembrane transport
transporter activity
ATP binding
sodium-exporting ATPase activity, phosphorylative mechanism
canalicular bile acid transmembrane transporter activity
bile acid-exporting ATPase activity
bile acid biosynthetic process
transport
bile acid and bile salt transport
canalicular bile acid transport
sodium ion transmembrane transport
transporter activity
ATP binding
sodium-exporting ATPase activity, phosphorylative mechanism
canalicular bile acid transmembrane transporter activity
bile acid-exporting ATPase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02039721.hg.1 | TC02002508.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02039721.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02039721.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000439188.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | ENST00000478354.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014848.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200208355.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200208355.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200208355.hg.1 | ENST00000263817.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200208355.hg.1 | NM_003742.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200208355.hg.1 | TC0200014848.hg.1 | Clariom D TC |
