Assay Details
Gene Symbol: SSBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256510.1 | NP_001243439.1 | 1 - 2 | 389 | 101 | |
GenBank mRNA | BC000895.1 | - | 1 - 2 | 389 | 3464282 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC000895.1, NM_001256510.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007319.hg.1 | ENST00000445848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007319.hg.1 | ENST00000445848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007319.hg.1 | ENST00000445848.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000265304.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000461433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000461433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000461433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000461433.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000463093.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000465167.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000467681.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000468267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000468267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000468267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000468267.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000469123.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000473783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000473783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000473783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000473783.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000481508.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000484178.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000489378.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000496622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000496622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000496622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000496622.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000498107.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | ENST00000612337.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256510.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256511.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_001256513.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NM_003143.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013469.hg.1 | NR_046269.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007319.hg.1 | ENST00000445848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007319.hg.1 | ENST00000445848.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007319.hg.1 | ENST00000445848.2 | Ensembl |
