Assay Details
| Gene Symbol: CARNS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011545191.2 | XP_011543493.1 | 1 | 1028 | 119 | |
| XM_017018054.1 | XP_016873543.1 | 2 | 730 | 119 | ||
| XM_017018055.1 | XP_016873544.1 | 1 | 100 | 119 | ||
| GenBank mRNA | AB037815.1 | - | 1 | 624 | 119 | |
| AK295331.1 | - | 2 | 730 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB037815.1, AK295331.1, XM_011545191.2, XM_017018054.1, XM_017018055.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000307823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000445895.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | ENST00000531958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_001166222.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013056.hg.1 | NM_020811.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232601 | - | - |

CARNS1
Genomic Map