Assay Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030930.3 | NP_112192.2 | 5 - 6 | 768 | 94 | |
| XM_011545290.1 | XP_011543592.1 | 3 - 4 | 378 | 94 | ||
| XM_011545291.2 | XP_011543593.1 | 4 - 5 | 660 | 94 | ||
| GenBank mRNA | AJ271326.1 | - | 5 - 6 | 727 | 94 | |
| AJ422142.1 | - | 5 - 6 | 736 | 94 | ||
| AY007125.1 | - | 4 - 5 | 555 | 94 | ||
| BC025669.1 | - | 5 - 6 | 714 | 94 | ||
| BC033623.1 | - | 5 - 6 | 766 | 94 | ||
| BC092472.1 | - | 5 - 6 | 736 | 30348121 | 94 | |
| BC101568.1 | - | 5 - 6 | 712 | 8069074 | 94 | |
| BC105104.1 | - | 5 - 6 | 712 | 8144107 | 94 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001999.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11023999.hg.1 | ENST00000227471.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023999.hg.1 | ENST00000530331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023999.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11023999.hg.1 | TC11001999.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000227471.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000524455.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000528423.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000533424.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000610659.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | ENST00000622364.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | NM_030930.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100187647.hg.1 | TC1100011367.hg.1 | Clariom D TC |

UNC93B1
Genomic Map