Assay Details
Gene Symbol: UNC93B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030930.3 | NP_112192.2 | 6 - 7 | 862 | 70 | |
XM_011545290.1 | XP_011543592.1 | 4 - 5 | 472 | 70 | ||
XM_011545291.2 | XP_011543593.1 | 5 - 6 | 754 | 70 | ||
GenBank mRNA | AJ271326.1 | - | 6 - 7 | 821 | 70 | |
AJ422142.1 | - | 6 - 7 | 830 | 70 | ||
AY007125.1 | - | 5 - 6 | 649 | 70 | ||
BC025669.1 | - | 6 - 7 | 808 | 70 | ||
BC033623.1 | - | 6 - 7 | 860 | 70 | ||
BC092472.1 | - | 6 - 7 | 830 | 30348121 | 70 | |
BC101568.1 | - | 6 - 7 | 806 | 8069074 | 70 | |
BC105104.1 | - | 6 - 7 | 806 | 8144107 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001999.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11023994.hg.1 | ENST00000227471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023994.hg.1 | ENST00000530331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023994.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023994.hg.1 | TC11001999.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11023998.hg.1 | ENST00000227471.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023998.hg.1 | ENST00000530331.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11023998.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11023998.hg.1 | TC11001999.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187644.hg.1 | TC1100011367.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000524455.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000525368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528096.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000528423.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000530138.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000531152.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000533424.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000610659.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | ENST00000622364.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100011367.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187639.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187639.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187639.hg.1 | ENST00000227471.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100187639.hg.1 | NM_030930.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100187639.hg.1 | TC1100011367.hg.1 | Clariom D TC |
