Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
ENST00000311191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
ENST00000389797.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
ENST00000389798.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
NM_001164211.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
NM_001164213.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
NM_015116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002121.hg.1 |
TC13000184.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
ENST00000311191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
ENST00000389797.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
ENST00000389798.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
NM_001164211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
NM_001164213.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
NM_015116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
ENST00000311191.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
ENST00000389797.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
ENST00000389798.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
NM_001164211.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
NM_001164213.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
NM_015116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148196.hg.1 |
TC1300007104.hg.1 |
Clariom D TC |