Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000342.hg.1 |
TC0X000025.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000342.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000342.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011259.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177182.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177182.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177182.hg.1 |
ENST00000217961.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177182.hg.1 |
NM_000351.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177182.hg.1 |
TC0X00011259.hg.1 |
Clariom D TC |