Assay Details
| Gene Symbol: METAP1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322278.1 | NP_001309207.1 | 7 - 8 | 962 | 92 | |
| NM_001322279.1 | NP_001309208.1 | 7 - 8 | 926 | 92 | ||
| NM_199227.2 | NP_954697.1 | 7 - 8 | 876 | 92 | ||
| NR_136273.1 | 6 - 7 | 813 | 92 | |||
| NR_136276.1 | 6 - 7 | 727 | 92 | |||
| XM_017003750.1 | XP_016859239.1 | 7 - 8 | 875 | 92 | ||
| GenBank mRNA | AY374142.1 | - | 7 - 8 | 876 | 92 | |
| BC113644.1 | - | 7 - 8 | 821 | 8322696 | 92 | |
| BC143913.1 | - | 7 - 8 | 907 | 92 | ||
| DQ005576.1 | - | 7 - 8 | 1190 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001025.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02015245.hg.1 | TC02001025.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015245.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015245.hg.1 | ENST00000488581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02015245.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000392582.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000491440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | ENST00000493742.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009966.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | ENST00000315796.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | ENST00000468537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | ENST00000488581.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | NM_199227.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200169918.hg.1 | TC0200009966.hg.1 | Clariom D TC |

METAP1D
Genomic Map