Assay Details
| Gene Symbol: BMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033403.1 | 4 - 5 | 817 | 102 | ||
| NR_033404.1 | 4 - 5 | 817 | 102 | |||
| GenBank mRNA | BC143338.1 | - | 4 - 5 | 622 | 102 | |
| BC144365.1 | - | 4 - 5 | 629 | 102 | ||
| BC144366.1 | - | 4 - 5 | 633 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

BMP1
Genomic Map