Assay Details
Gene Symbol: CALY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321617.1 | NP_001308546.1 | 3 - 4 | 415 | 76 | |
NM_015722.3 | NP_056537.1 | 3 - 4 | 403 | 76 | ||
GenBank mRNA | AF225903.1 | - | 3 - 4 | 340 | 76 | |
BC038978.1 | - | 3 - 4 | 304 | 5734358 | 76 | |
BI914942.1 | - | 3 - 4 | 403 | 76 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001787.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001321617.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | ENST00000605518.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023607.hg.1 | ENST00000252939.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023607.hg.1 | ENST00000368556.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023607.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023607.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023607.hg.1 | TC10001787.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10023609.hg.1 | ENST00000252939.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023609.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023609.hg.1 | ENST00000368556.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023609.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10023609.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10023609.hg.1 | TC10001787.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184613.hg.1 | TC1000012395.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368555.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000467611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | ENST00000605518.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000012395.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | ENST00000252939.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | ENST00000368558.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | NM_015722.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000184611.hg.1 | TC1000012395.hg.1 | Clariom D TC |
