Assay Details
Gene Symbol: WAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000377.2 | NP_000368.1 | 10 - 11 | 1392 | 106 | |
XM_011543977.2 | XP_011542279.1 | 10 - 11 | 1241 | 106 | ||
XM_017029786.1 | XP_016885275.1 | 10 - 11 | 1436 | 106 | ||
GenBank mRNA | BC002961.1 | - | 10 - 11 | 1371 | 106 | |
BC012738.2 | - | 10 - 11 | 1363 | 3688197 | 106 | |
U12707.1 | - | 10 - 11 | 1369 | 106 | ||
U19927.1 | - | 10 - 11 | 1359 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000257.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of T cell antigen processing and presentation
protein complex assembly
defense response
immune response
blood coagulation
actin polymerization or depolymerization
epidermis development
endosomal transport
actin filament polymerization
actin filament-based movement
Fc-gamma receptor signaling pathway involved in phagocytosis
T cell activation
regulation of catalytic activity
T cell receptor signaling pathway
negative regulation of cell motility
positive regulation of Arp2/3 complex-mediated actin nucleation
actin binding
protein binding
SH3 domain binding
protein kinase binding
GTPase regulator activity
identical protein binding
phospholipase binding
regulation of T cell antigen processing and presentation
protein complex assembly
defense response
immune response
blood coagulation
actin polymerization or depolymerization
epidermis development
endosomal transport
actin filament polymerization
actin filament-based movement
Fc-gamma receptor signaling pathway involved in phagocytosis
T cell activation
regulation of catalytic activity
T cell receptor signaling pathway
negative regulation of cell motility
positive regulation of Arp2/3 complex-mediated actin nucleation
actin binding
protein binding
SH3 domain binding
protein kinase binding
GTPase regulator activity
identical protein binding
phospholipase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003171.hg.1 | TC0X000257.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003171.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003171.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003172.hg.1 | TC0X000257.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003172.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X003172.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149226.hg.1 | TC0X00007199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00149225.hg.1 | TC0X00007199.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000376701.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000450772.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000465982.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000470107.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000474174.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000483750.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | ENST00000490627.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007199.hg.1 | NM_000377.2 | Refseq |
