Assay Details
| Gene Symbol: PUSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153339.1 | NP_699170.1 | 2 - 3 | 166 | 109 | |
| XM_005244720.3 | XP_005244777.1 | 2 - 3 | 218 | 109 | ||
| GenBank mRNA | AK075292.1 | - | 2 - 3 | 183 | 109 | |
| BC034304.1 | - | 2 - 3 | 166 | 4778177 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000033.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01000408.hg.1 | TC01000033.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000408.hg.1 | ENST00000379031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000408.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144733.hg.1 | TC0100006499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |

PUSL1
Genomic Map