Assay Details
Gene Symbol: GSK3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019884.2 | NP_063937.2 | 3 - 4 | 677 | 90 | |
GenBank mRNA | BC027984.1 | - | 3 - 4 | 696 | 3903896 | 90 |
BC051865.2 | - | 3 - 4 | 666 | 6171438 | 90 | |
CR749620.1 | - | 2 - 3 | 2025 | 90 | ||
D63424.1 | - | 3 - 4 | 649 | 90 | ||
L40027.1 | - | 3 - 4 | 672 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001569.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
glycogen metabolic process
protein phosphorylation
dopamine receptor signaling pathway
nervous system development
insulin receptor signaling pathway
negative regulation of UDP-glucose catabolic process
Wnt signaling pathway
positive regulation of cAMP biosynthetic process
negative regulation of TOR signaling
cellular response to insulin stimulus
cellular response to interleukin-3
IRE1-mediated unfolded protein response
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of glycogen biosynthetic process
positive regulation of protein catabolic process
positive regulation of heart contraction
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of cell growth involved in cardiac muscle cell development
positive regulation of adrenergic receptor signaling pathway
negative regulation of canonical Wnt signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
chemical synaptic transmission, postsynaptic
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
negative regulation of glycogen synthase activity, transferring glucose-1-phosphate
negative regulation of type B pancreatic cell development
negative regulation of glycogen (starch) synthase activity
positive regulation of glycogen (starch) synthase activity
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase A catalytic subunit binding
tau-protein kinase activity
regulation of systemic arterial blood pressure
cardiac left ventricle morphogenesis
glycogen metabolic process
protein phosphorylation
dopamine receptor signaling pathway
nervous system development
insulin receptor signaling pathway
negative regulation of UDP-glucose catabolic process
Wnt signaling pathway
positive regulation of cAMP biosynthetic process
negative regulation of TOR signaling
cellular response to insulin stimulus
cellular response to interleukin-3
IRE1-mediated unfolded protein response
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of glycogen biosynthetic process
positive regulation of protein catabolic process
positive regulation of heart contraction
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of cell growth involved in cardiac muscle cell development
positive regulation of adrenergic receptor signaling pathway
negative regulation of canonical Wnt signaling pathway
extrinsic apoptotic signaling pathway in absence of ligand
chemical synaptic transmission, postsynaptic
positive regulation of mitochondrial outer membrane permeabilization involved in apoptotic signaling pathway
regulation of mitophagy
positive regulation of protein targeting to mitochondrion
negative regulation of glycogen synthase activity, transferring glucose-1-phosphate
negative regulation of type B pancreatic cell development
negative regulation of glycogen (starch) synthase activity
positive regulation of glycogen (starch) synthase activity
protein serine/threonine kinase activity
protein binding
ATP binding
protein kinase A catalytic subunit binding
tau-protein kinase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR19021400.hg.1 | ENST00000222330.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19021400.hg.1 | ENST00000398249.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19021400.hg.1 | ENST00000544315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19021400.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19021400.hg.1 | TC19001569.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | ENST00000493059.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011957.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | NM_019884.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | TC1900011957.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | ENST00000222330.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | ENST00000398249.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | ENST00000453535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900206333.hg.1 | NM_019884.2 | Refseq |
