Assay Details
Gene Symbol: MAP4K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006575.4 | NP_006566.2 | 22 - 23 | 1964 | 142 | |
NM_198794.2 | NP_942089.1 | 22 - 23 | 2012 | 142 | ||
XM_006720013.3 | XP_006720076.1 | 22 - 23 | 1859 | 142 | ||
XM_011536377.2 | XP_011534679.1 | 22 - 23 | 1873 | 142 | ||
XM_011536378.2 | XP_011534680.1 | 22 - 23 | 1709 | 142 | ||
XM_011536379.2 | XP_011534681.1 | 18 - 19 | 1820 | 142 | ||
XM_011536380.2 | XP_011534682.1 | 9 - 10 | 2606 | 142 | ||
GenBank mRNA | AK124738.1 | - | 8 - 9 | 2397 | 142 | |
AK313612.1 | - | 21 - 22 | 1770 | 142 | ||
BC036013.1 | - | 22 - 23 | 1902 | 5272190 | 142 | |
U77129.1 | - | 22 - 23 | 1964 | 142 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14001120.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKKK activity
protein phosphorylation
activation of JUN kinase activity
intracellular signal transduction
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
MAP kinase kinase kinase kinase activity
activation of MAPKKK activity
protein phosphorylation
activation of JUN kinase activity
intracellular signal transduction
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
MAP kinase kinase kinase kinase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000013125.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000364703.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554066.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554091.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554273.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000554990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000555216.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557124.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557124.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557124.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557124.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557210.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557390.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | ENST00000557578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_006575.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009131.hg.1 | NM_198794.2 | Refseq |
