Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009975.hg.1 |
ENST00000263674.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009975.hg.1 |
NM_014786.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11009975.hg.1 |
TC11000774.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
ENST00000263674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
ENST00000543530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
NM_014786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
ENST00000263674.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
ENST00000543530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
NM_014786.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100162000.hg.1 |
TC1100008382.hg.1 |
Clariom D TC |