Assay Details
Gene Symbol: ARHGEF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014786.3 | NP_055601.2 | 19 - 20 | 5743 | 75 | |
XM_017018623.1 | XP_016874112.1 | 19 - 20 | 4134 | 75 | ||
XM_017018624.1 | XP_016874113.1 | 18 - 19 | 3213 | 75 | ||
GenBank mRNA | AB002335.1 | - | 19 - 20 | 4517 | 75 | |
AF378754.1 | - | 19 - 20 | 5737 | 75 | ||
BC137228.1 | - | 19 - 20 | 5743 | 9021225 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000774.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
Function(s)
Process(es)
actin cytoskeleton organization
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
actin cytoskeleton organization
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11009976.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11009976.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11009976.hg.1 | TC11000774.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000536481.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000537198.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | ENST00000544519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100008382.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | ENST00000263674.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | ENST00000543530.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | NM_014786.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100162001.hg.1 | TC1100008382.hg.1 | Clariom D TC |
