Assay Details
Gene Symbol: PARVA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018222.4 | NP_060692.2 | 9 - 10 | 969 | 74 | |
XM_005253015.3 | XP_005253072.1 | 9 - 10 | 958 | 74 | ||
GenBank mRNA | AF237771.1 | - | 9 - 10 | 892 | 74 | |
AF325830.1 | - | 9 - 10 | 800 | 74 | ||
AK001655.1 | - | 9 - 10 | 849 | 74 | ||
AK098504.1 | - | 8 - 9 | 893 | 74 | ||
BC014535.1 | - | 9 - 10 | 828 | 4065820 | 74 | |
BC016713.1 | - | 9 - 10 | 886 | 4065758 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000201.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
outflow tract septum morphogenesis
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein binding
cadherin binding involved in cell-cell adhesion
sprouting angiogenesis
outflow tract septum morphogenesis
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11002714.hg.1 | ENST00000334956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11002714.hg.1 | ENST00000539723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11002714.hg.1 | ENST00000550549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11002714.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11002714.hg.1 | TC11000201.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11002715.hg.1 | ENST00000334956.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11002715.hg.1 | ENST00000539723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11002715.hg.1 | ENST00000550549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11002715.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11002715.hg.1 | TC11000201.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100148456.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100148456.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100148456.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100148456.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100148456.hg.1 | TC1100006879.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100148457.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100148457.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100148457.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100148457.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100148457.hg.1 | TC1100006879.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000334956.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000526746.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000528916.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000530755.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533345.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | ENST00000533392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006879.hg.1 | NM_018222.4 | Refseq |
