Assay Details
Gene Symbol: OBFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024928.4 | NP_079204.2 | 5 - 6 | 714 | 86 | |
XM_006717976.3 | XP_006718039.1 | 5 - 6 | 714 | 86 | ||
XM_011540184.2 | XP_011538486.1 | 5 - 6 | 714 | 86 | ||
XM_017016669.1 | XP_016872158.1 | 4 - 5 | 881 | 86 | ||
XM_017016670.1 | XP_016872159.1 | 5 - 6 | 714 | 86 | ||
GenBank mRNA | AK026212.1 | - | 4 - 5 | 539 | 86 | |
BC017400.2 | - | 5 - 6 | 635 | 3029792 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001636.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
telomere maintenance
telomere maintenance via telomere lengthening
telomere capping
positive regulation of telomere maintenance via telomerase
positive regulation of DNA replication
positive regulation of telomerase activity
negative regulation of telomerase activity
regulation of telomere maintenance via telomere lengthening
single-stranded DNA binding
protein binding
telomerase inhibitor activity
telomeric DNA binding
single-stranded telomeric DNA binding
telomere maintenance
telomere maintenance via telomere lengthening
telomere capping
positive regulation of telomere maintenance via telomerase
positive regulation of DNA replication
positive regulation of telomerase activity
negative regulation of telomerase activity
regulation of telomere maintenance via telomere lengthening
single-stranded DNA binding
protein binding
telomerase inhibitor activity
telomeric DNA binding
single-stranded telomeric DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10021399.hg.1 | ENST00000224950.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021399.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021399.hg.1 | ENST00000466828.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021399.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10021399.hg.1 | TC10001636.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000008001.hg.1 | ENST00000431638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000008001.hg.1 | ENST00000431638.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021401.hg.1 | ENST00000224950.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021401.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021401.hg.1 | ENST00000466828.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021401.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10021401.hg.1 | TC10001636.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000568017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000568017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | ENST00000568017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000180316.hg.1 | TC1000011736.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000180318.hg.1 | TC1000011736.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000008001.hg.1 | ENST00000431638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000008001.hg.1 | ENST00000431638.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000224950.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000369764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000466828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000472951.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000568017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000568017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | ENST00000568017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011736.hg.1 | NM_024928.4 | Refseq |
