Assay Details
| Gene Symbol: GLS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280796.1 | NP_001267725.1 | 4 - 5 | 835 | 95 | |
| NM_013267.3 | NP_037399.2 | 4 - 5 | 835 | 95 | ||
| XM_005268797.1 | XP_005268854.1 | 4 - 5 | 462 | 95 | ||
| GenBank mRNA | AF110330.1 | - | 4 - 5 | 590 | 95 | |
| AF223944.1 | - | 4 - 5 | 762 | 95 | ||
| AK000467.1 | - | 4 - 5 | 582 | 95 | ||
| AK294822.1 | - | 4 - 5 | 780 | 95 | ||
| AK303772.1 | - | 4 - 5 | 636 | 95 | ||
| BC119767.1 | - | 4 - 5 | 780 | 95 | ||
| BC119768.1 | - | 4 - 5 | 780 | 95 | ||
| HM775422.1 | - | 3 - 4 | 454 | 95 | ||
| HM775424.1 | - | 3 - 4 | 454 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001280796.1 transcripts.
Set Membership: |
|

GLS2
Genomic Map