Assay Details
| Gene Symbol: RBM15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB209680.1 | - | 1 | 1016 | 183 | |
| AF368064.1 | - | 1 | 3022 | 183 | ||
| AK025596.1 | - | 1 | 1687 | 183 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000369784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000487146.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000602849.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000617047.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | ENST00000618772.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_001201545.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009383.hg.1 | NM_022768.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 102051 | - | - |

RBM15
Genomic Map