Assay Details
| Gene Symbol: CRTAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019604.3 | NP_062550.2 | 2 - 3 | 245 | 76 | |
| GenBank mRNA | AF001622.1 | - | 2 - 3 | 196 | 76 | |
| AK313578.1 | - | 2 - 3 | 243 | 76 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001116.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
T cell mediated cytotoxicity
detection of tumor cell
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
regulation of immune response
activated T cell proliferation
detection of stimulus
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding
T cell mediated cytotoxicity
detection of tumor cell
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
positive regulation of natural killer cell mediated cytotoxicity
positive regulation of cytokine secretion
regulation of immune response
activated T cell proliferation
detection of stimulus
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11014221.hg.1 | ENST00000227348.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014221.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014221.hg.1 | TC11001116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014218.hg.1 | ENST00000227348.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014218.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014218.hg.1 | TC11001116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014219.hg.1 | ENST00000227348.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014219.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014219.hg.1 | TC11001116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014220.hg.1 | ENST00000227348.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014220.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11014220.hg.1 | TC11001116.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170054.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170054.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170054.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170054.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170054.hg.1 | TC1100009334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | ENST00000533709.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_001304782.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009334.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170056.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170056.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170056.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170056.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170056.hg.1 | TC1100009334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100170057.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170057.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170057.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170057.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170057.hg.1 | TC1100009334.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100170055.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170055.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170055.hg.1 | ENST00000227348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100170055.hg.1 | NM_019604.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100170055.hg.1 | TC1100009334.hg.1 | Clariom D TC |

CRTAM
Genomic Map