| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000008.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000008.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000008.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000008.hg.1 |
TC20000002.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000007.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000007.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000007.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20000007.hg.1 |
TC20000002.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000006434.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144051.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144051.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144051.hg.1 |
TC2000006434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000144051.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144051.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
NM_030931.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
TC2000006434.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
ENST00000382398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
ENST00000542572.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000144053.hg.1 |
NM_030931.3 |
Refseq |